Learn More
Description
- Assay Advantages: simple measurement of deacetylating activities
- Highly sensitive, fast data acquisition
Specifications
Specifications
| Quantity | 10 mL |
| Content And Storage | -20°C |
| For Use With (Application) | Determining SIRT inhibitor or activator potency, SIRT Enzyme profiling |
| Kit Contents | 1 x 1 Vial SIRT-Glo substrate, 1 x 10μL developer reagent, 1 x 30μL Nicotinamide, 1 x 10μL HeLa nuclear extract, 1 x 25mL SIRT-Glo Buffer |
| pH | 8 |
| Product Type | Screening System |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.