Learn More
SIRT1 Mouse anti-Human, Mouse, Clone: 7F6G6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SIRT1 |
---|---|
Clone | 7F6G6 |
Concentration | 1.2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6396
![]() ![]() |
Proteintech Group Inc
603031IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human SIRT1.
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Alternative splicing results in multiple transcript variants.Specifications
SIRT1 | |
1.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hSIR2, hSIRT1, SIR2 like protein 1, SIR2L1, SIRT1 | |
Sirt1 | |
IgG2b | |
Protein A | |
Antibody |
7F6G6 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q923E4, Q96EB6 | |
23411, 93759 | |
SIRT1 Fusion Protein Ag17677 | |
Primary | |
-20°C | |
SIRT1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.