Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117095 Supplier Invitrogen™ Supplier No. PA5117095
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, 22Rv1, SW620, MCF7 Immunogen sequence: QRETGKLDGP DK.

SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The SIRT3 is included in class I of the sirtuin family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT3
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SIRT3
Gene Accession No. Q8R104, Q9NTG7
Gene Alias 2310003L23Rik; AI848213; HGNC:14931; hSIRT3; mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase; mitochondrial protein lysine deacetylase; mSIR2L3; NAD-dependent deacetylase sirtuin-3; NAD-dependent deacetylase sirtuin-3, mitochondrial; NAD-dependent protein deacetylase sirtuin-3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; regulatory protein SIR2 homolog 3; silent mating type information regulation 2, (S.cerevisiae, homolog)-like 3; silent mating type information regulation 2, S.cerevisiae, homolog 3; Sir2l3; sir2-like 3; SIR2-like protein 3; SIRT3; SIRT3L mitochondrial; sirtuin 3; sirtuin type 3
Gene Symbols SIRT3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23410, 293615, 64384
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.