Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT3 Polyclonal Antibody

SIRT3 Polyclonal Antibody
Supplier: Thermo Scientific PA520971
Description
SIRT3 Polyclonal Antibody for Western Blot
Specifications
| SIRT3 | |
| Polyclonal | |
| Unconjugated | |
| SIRT3 | |
| 2310003L23Rik; AI848213; HGNC:14931; hSIRT3; mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase; mitochondrial protein lysine deacetylase; mSIR2L3; NAD-dependent deacetylase sirtuin-3; NAD-dependent deacetylase sirtuin-3, mitochondrial; NAD-dependent protein deacetylase sirtuin-3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; regulatory protein SIR2 homolog 3; silent mating type information regulation 2, (S.cerevisiae, homolog)-like 3; silent mating type information regulation 2, S.cerevisiae, homolog 3; Sir2l3; sir2-like 3; SIR2-like protein 3; SIRT3; SIRT3L mitochondrial; sirtuin 3; sirtuin type 3 | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| 23410, 293615, 64384 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8R104, Q9NTG7 | |
| SIRT3 | |
| A 17 amino acid peptide near the center of human SIRT3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction