Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT3 Rabbit anti-Chicken, Human, Mouse, Non-human Primate, Rat, Biotin, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc SIR3BIOTIN

Description
SIRT3 Polyclonal antibody specifically detects SIRT3 in Chicken, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
SIRT3 | |
Polyclonal | |
Biotin | |
SIRT3 | |
2310003L23Rik; AI848213; HGNC:14931; hSIRT3; mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase; mitochondrial protein lysine deacetylase; mSIR2L3; NAD-dependent deacetylase sirtuin-3; NAD-dependent deacetylase sirtuin-3, mitochondrial; NAD-dependent protein deacetylase sirtuin-3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; regulatory protein SIR2 homolog 3; silent mating type information regulation 2, (S.cerevisiae, homolog)-like 3; silent mating type information regulation 2, S.cerevisiae, homolog 3; Sir2l3; sir2-like 3; SIR2-like protein 3; SIRT3; SIRT3L mitochondrial; sirtuin 3; sirtuin type 3 | |
Synthetic peptide:RDLVGPFVLSPRRKDVVQLcorresponding to amino acids 203-221 of rat SIRT3 protein. | |
200 μL | |
Primary | |
Chicken, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q8R104, Q9NTG7 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23410, 293615, 422988, 450911, 64384 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction