Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT4 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298856R
Description
SIRT4 Polyclonal specifically detects SIRT4 in Human samples. It is validated for Western Blot.Specifications
SIRT4 | |
Polyclonal | |
Western Blot | |
EC 2.4.2.-, EC 3.5.1, MGC130046, MGC130047, MGC57437, SIR2L4NAD-dependent ADP-ribosyltransferase sirtuin-4, sir2-like 4, SIR2-like protein 4, sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 4, sirtuin 4, sirtuin type 4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SIRT4. | |
0.1 mL | |
DNA Repair, Epigenetics, Homologous Recombination | |
23409 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction