Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987814
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7814 Supplier Bioss Supplier No. BS7537R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class IV of the sirtuin family.

Specifications

Antigen SIRT4
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SIRT4
Gene Accession No. Q9Y6E7
Gene Alias 4930596O17Rik; BOS_16541; LOC100353055; MGC130046; MGC130047; MGC57437; NAD-dependent ADP-ribosyltransferase sirtuin-4; NAD-dependent deacetylase sirtuin-4; NAD-dependent protein deacetylase sirtuin-4; NAD-dependent protein lipoamidase sirtuin-4, mitochondrial; Regulatory protein SIR2 homolog 4; Sir2l4; sir2-like 4; SIR2-like protein 4; SIRT4; sirtuin 4; sirtuin type 4
Gene Symbols SIRT4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SIRT4/SIR 2 like protein 4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23409
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.