Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT5 Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503233S


Description
SIRT5 Monoclonal antibody specifically detects SIRT5 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
SIRT5 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q68FX9, Q9NXA8 | |
Mouse | |
Affinity chromatography | |
RUO | |
23408, 306840 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2C4 | |
Unconjugated | |
SIRT5 | |
0610012J09Rik; 1500032M05Rik; AV001953; FLJ36950; HAMAP-Rule:MF_03160}; hypothetical protein LOC507347; NAD-dependent deacetylase sirtuin-5; NAD-dependent deacetylase sirtuin-5-like protein; NAD-dependent lysine demalonylase and desuccinylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial {ECO:0000255; Regulatory protein SIR2 homolog 5; regulatory protein SIR2 homolog 5 {ECO:0000255; si:ch211-121a2.1; silent mating type information regulation 2, S.cerevisiae, homolog 5; Sir2l5; sir2-like 5; SIR2-like protein 5; SIR2-like protein 5 {ECO:0000255; Sirt5; sirtuin; sirtuin 5; sirtuin type 5; zgc:92288 | |
Full length human recombinant protein of human SIRT5 produced in HEK293 cell. | |
30 μL | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction