Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540535 Supplier Invitrogen™ Supplier No. PA540535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EVDRELAHCD LCLVVGTSSV VYPAAMFAPQ VAARGVPVAE FNTETTPATN Sequence homology: Cow: 93%; Dog: 93%; Goat: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%; Zebrafish: 100%.

SIRT5 is a human member of a family of proteins called Sirtuins (Sir2-like proteins) and are present in prokaryotes and eukaryotes. All Sir2-like proteins have a sirtuin core domain, which contains a series of sequence motifs conserved in organisms ranging from bacteria to humans. Bacterial, yeast and mammalian sirtuins are able to metabolize NAD and possibly at as mono-ADP-ribosyltransferases. The enzymatic function of sirtuins is not yet completely understood but recent reports of histone-activated Sir2-mediated NAD metabolism and NAD-activated Sir2-mediated histone deacetylation suggest a possible coupled reciprocal activation mechanism involving interactions of Sir2 with NAD and the N epsilon-acetyl-lysine groups of acetylated histones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SIRT5
Gene Accession No. Q9NXA8
Gene Alias 0610012J09Rik; 1500032M05Rik; AV001953; FLJ36950; HAMAP-Rule:MF_03160}; hypothetical protein LOC507347; NAD-dependent deacetylase sirtuin-5; NAD-dependent deacetylase sirtuin-5-like protein; NAD-dependent lysine demalonylase and desuccinylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial {ECO:0000255; Regulatory protein SIR2 homolog 5; regulatory protein SIR2 homolog 5 {ECO:0000255; si:ch211-121a2.1; silent mating type information regulation 2, S.cerevisiae, homolog 5; Sir2l5; sir2-like 5; SIR2-like protein 5; SIR2-like protein 5 {ECO:0000255; Sirt5; sirtuin; sirtuin 5; sirtuin type 5; zgc:92288
Gene Symbols SIRT5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SIRT5 (aa 234-283).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23408
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.