Learn More
Invitrogen™ SIRT5 Recombinant Mouse Monoclonal Antibody, Biotin
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ 730092

Description
730092 is a biotinylated Fab fragment targeting SIRT5. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
SIRT5 is a human member of a family of proteins called Sirtuins (Sir2-like proteins) and are present in prokaryotes and eukaryotes. All Sir2-like proteins have a sirtuin core domain, which contains a series of sequence motifs conserved in organisms ranging from bacteria to humans. Bacterial, yeast and mammalian sirtuins are able to metabolize NAD and possibly at as mono-ADP-ribosyltransferases. The enzymatic function of sirtuins is not yet completely understood but recent reports of histone-activated Sir2-mediated NAD metabolism and NAD-activated Sir2-mediated histone deacetylation suggest a possible coupled reciprocal activation mechanism involving interactions of Sir2 with NAD and the N epsilon-acetyl-lysine groups of acetylated histones.
Specifications
SIRT5 | |
Recombinant Monoclonal | |
Biotin | |
SIRT5 | |
0610012J09Rik; 1500032M05Rik; AV001953; FLJ36950; HAMAP-Rule:MF_03160}; hypothetical protein LOC507347; NAD-dependent deacetylase sirtuin-5; NAD-dependent deacetylase sirtuin-5-like protein; NAD-dependent lysine demalonylase and desuccinylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial; NAD-dependent protein deacylase sirtuin-5, mitochondrial {ECO:0000255; Regulatory protein SIR2 homolog 5; regulatory protein SIR2 homolog 5 {ECO:0000255; si:ch211-121a2.1; silent mating type information regulation 2, S.cerevisiae, homolog 5; Sir2l5; sir2-like 5; SIR2-like protein 5; SIR2-like protein 5 {ECO:0000255; Sirt5; sirtuin; sirtuin 5; sirtuin type 5; zgc:92288 | |
Mouse | |
Protein A | |
RUO | |
23408 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Immunoprecipitation | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9NXA8 | |
SIRT5 | |
34-302 of 310 of SIRT5 Protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.