Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002524F
Description
SIRT6 Polyclonal specifically detects SIRT6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| SIRT6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| Q8N6T7 | |
| SIRT6 | |
| A synthetic peptide made to a C-terminal region of the human SIRT6 protein (within residues 300-355). [Swiss-Prot Q8N6T7] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 72% homology to rat. | |
| Store at 4C in the dark. |
| Immunocytochemistry, Immunofluorescence | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Cancer, Chromatin Modifiers, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific, Signal Transduction | |
| 51548 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction