Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$192.47 - $529.95
Specifications
| Antigen | SIRT6 |
|---|---|
| Concentration | 1.05 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002524SS
![]() |
Novus Biologicals
NB1002524SS |
0.025 mL |
Each for $192.47
|
|
|||||
NB1002524
![]() |
Novus Biologicals
NB1002524 |
0.1 mL |
Each for $529.95
|
|
|||||
Description
SIRT6 Polyclonal specifically detects SIRT6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| SIRT6 | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Chromatin Modifiers, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Signal Transduction | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
| SIRT6 | |
| IgG | |
| Affinity Purified | |
| Human. Immunogen sequence has 72% homology to rat. |
| 1.05 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8N6T7 | |
| 51548 | |
| A synthetic peptide made to a C-terminal region of the human SIRT6 protein (within residues 300-355). [Swiss-Prot Q8N6T7] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 39 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title