Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Antibody (OTI1G3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201310
Description
SIRT6 Monoclonal specifically detects SIRT6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SIRT6 | |
Monoclonal | |
0.6 mg/mL | |
Western Blot 1:500-2000, Immunocytochemistry/Immunofluorescence 1:100 | |
SIRT6 | |
Human recombinant protein fragment corresponding to amino acids 88-352 of human SIRT6(NP_057623) produced in E.coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1G3 | |
Unconjugated | |
EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
Mouse | |
38.9 kDa | |
0.1 mL | |
Cancer, Chromatin Modifiers, Chromatin Research, DNA Repair, Epigenetics, Histone Deacetylases, Signal Transduction | |
51548 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction