Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Mouse anti-Human, Clone:2G1H1, Abnova™
Mouse monoclonal antibody raised against recombinant human SIRT6.
Supplier: Abnova Corporation MAB17126
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class IV of the sirtuin family. [provided by RefSeq]Specifications
SIRT6 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
SIRT6 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
2G1H1 | |
sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae) | |
In PBS (0.05% sodium azide) | |
SIR2L6 | |
Recombinant protein corresponding to amino acid 141-250 of human SIRT6 from E. coli. | |
51548 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction