Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Mouse anti-Human, Clone: OTI1G3, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503670S

Description
The Silent Information Regulator (SIR2) family of genes are highly conserved from prokaryotes to eukaryotes and have important functions in the regulation of metabolism, growth and differentiation, inflammation, cellular survival, as well as in senescence and lifespan extension. Sirtuins, including SIRT1-7, are human homologs of yeast Sir2p. Sirtuins are NAD+-dependent histone/protein deacetylases (HDAC) which regulate cellular metabolism, e.g. energy metabolism, and thereby are associated with aging and several age-related diseases. SIRT6 is a histone H3 lysine 9 deacetylase and is thought to stabilize the DNA-dependent protein kinase at double-stranded DNA breaks and may have a role in the process of mammalian aging.Specifications
SIRT6 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N6T7 | |
SIRT6 | |
Human recombit protein fragment corresponding to amino acids 88-352 of human SIRT6 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1G3 | |
Unconjugated | |
SIRT6 | |
SIR2L6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51548 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction