Learn More
Invitrogen™ SIRT6 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-20967 can be used with blocking peptide PEP-1081.
Specifications
Specifications
| Antigen | SIRT6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SIRT6 |
| Gene Accession No. | P59941, Q8N6T7 |
| Gene Alias | 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6 |
| Gene Symbols | SIRT6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.