Learn More
Invitrogen™ SIRT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595240
Description
Human SIRT6 shares 95% amino acid (aa) sequence identity with mouse SIRT6. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U87 whole cell, human A549 whole cell, human K562 whole cell, human U2OS whole cell, human Hela whole cell. IHC: Mouse Cardiac Muscle Tissue, Human Mammary Cancer Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
Specifications
| SIRT6 | |
| Polyclonal | |
| Unconjugated | |
| SIRT6 | |
| 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50721, 51548 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P59941, Q8N6T7 | |
| SIRT6 | |
| E.coli-derived human SIRT6 recombinant protein (Position: D14-E180). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.