Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT6 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992953
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2953 10 μL
50-199-2952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2953 Supplier Active Motif Supplier No. 39912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.

Specifications

Antigen SIRT6
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SIRT6
Gene Accession No. Q8N6T7
Gene Alias 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; SIRT6; sirtuin 6; sirtuin type 6
Gene Symbols SIRT6
Host Species Rabbit
Immunogen This SIRT6 antibody was raised against a peptide within the C-terminus of human SIRT6.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51548
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.