Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Rabbit anti-Human, Clone: 1D0L6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | SIRT6 |
---|---|
Clone | 1D0L6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SIRT6 Monoclonal antibody specifically detects SIRT6 in Human samples. It is validated for Western BlotSpecifications
SIRT6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, DNA Repair, Epigenetics, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
51548 | |
IgG | |
Affinity purified |
1D0L6 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
A synthetic peptide corresponding to a sequence within amino acids 256-355 of human SIRT6 (Q8N6T7). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title