Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SIRT6 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SIRT6 Polyclonal specifically detects SIRT6 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.Specifications
SIRT6 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 3.5.1.-, NAD-dependent deacetylase sirtuin-6, SIR2L6sir2-related protein type 6, SIR2-like protein 6, sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S. cerevisiae, homolog) 6, sirtuin 6, sirtuin type 6 | |
The immunogen is a synthetic peptide directed towards the middle region of human SIRT6 (NP_057623). Peptide sequence TRINGSIPAGPKQEPCAQHNGSEPASPKRERPTSPAPHRPPKRVKAKAVP | |
Affinity purified |
Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, DNA Repair, Epigenetics, Phospho Specific, Signal Transduction | |
PBS buffer, 2% sucrose | |
51548 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title