Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRT6 Recombinant Rabbit Monoclonal Antibody (JJ20-69)
GREENER_CHOICE

Catalog No. PIMA532437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532437 Supplier Invitrogen™ Supplier No. MA532437
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT6
Applications Western Blot
Classification Recombinant Monoclonal
Clone JJ20-69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene SIRT6
Gene Accession No. Q8N6T7
Gene Alias 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6
Gene Symbols SIRT6
Host Species Rabbit
Immunogen Synthetic peptide within Human SIRT6 aa 306-355.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51548
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.