Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRT7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $426.00
Specifications
Antigen | SIRT7 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056364T
![]() |
Novus Biologicals
NB10056364SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056364
![]() |
Novus Biologicals
NB10056364 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
SIRT7 Polyclonal specifically detects SIRT7 in Human samples. It is validated for Western Blot.Specifications
SIRT7 | |
Western Blot 1-3 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Q9NRC8 | |
51547 | |
A mixture of synthetic peptides corresponding to amino acids 35-51 and 361-377 of human SIRT7 was used as immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair | |
PBS with 0.05% Sodium Azide | |
EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7 | |
SIRT7 | |
IgG | |
Protein G purified | |
There is no corresponding 40 kD band seen in mouse or rat liver cell lysate. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title