Learn More
Invitrogen™ SIRT7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587543
Description
Immunogen sequence: NREYVRVFDV TERTALHRHQ TGRTCHKCGT QLRDTIVHFG ERGTLGQPLN WEAATEAASR ADTILCLGSS LKVLKKYPRL WCMTKPPSRR PKLYIVNLQW TPKDDWAALK LHGKCDDVMR LLMAELGLEI PAYSRWQDPI FSLATPLRAG EEGSHSRKSL CRSREEAPPG DRGAPLSSAP ILGGWFGRGC TKRTKRKKVT; Positive Samples: HepG2, SW480; Cellular Location: Cytoplasm, Nucleus, nucleolus.
SIRT7 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. SIRT7 is included in class IV. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. SIRT7 is included in class IV of the sirtuin family.
Specifications
| SIRT7 | |
| Polyclonal | |
| Unconjugated | |
| SIRT7 | |
| MGC126840; MGC126842; NAD-dependent deacetylase sirtuin-7; NAD-dependent protein deacetylase sirtuin-7; NAD-dependent protein deacylase sirtuin-7; regulatory protein SIR2 homolog 7; silent mating type information regulation 2, S.cerevisiae, homolog 7; Sir2l7; SIR2-like protein 7; sir2-related protein type 7; Sirt7; sirtuin 7; sirtuin type 7; Unknown (protein for MGC:142712) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 209011, 303745, 51547 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| B2RZ55, Q8BKJ9, Q9NRC8 | |
| SIRT7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 201-400 of human SIRT7 (NP_0576221). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.