Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody (012), Janelia Fluor™ 646 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289977JF646
Description
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for ELISASpecifications
Sirtuin 1/SIRT1 | |
Monoclonal | |
Janelia Fluor 646 | |
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Sirtuin 1/SIRT1 (NP_036370.2; Met 193-Ser 747). | |
0.1 mL | |
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific | |
23411 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
012 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction