Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody (1F3), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151641G
Description
Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Sirtuin 1/SIRT1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SIRT1 | |
Purified recombinant fragment of human SIRT1 (amino acids: 265-452) expressed in E. coli. [UniProt# Q96EB6] | |
0.1 mL | |
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific | |
23411 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1F3 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction