Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 1/SIRT1 Antibody (1F3), FITC, Novus Biologicals™
SDP

Catalog No. NBP151641F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151641F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151641F Supplier Novus Biologicals Supplier No. NBP151641F
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Sirtuin 1/SIRT1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1F3
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Gene Symbols SIRT1
Host Species Mouse
Immunogen Purified recombinant fragment of human SIRT1 (amino acids: 265-452) expressed in E. coli. [UniProt# Q96EB6]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Histone Deacetylases, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.