Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP149540
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.Specifications
| Sirtuin 1/SIRT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide | |
| EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:150, Knockout Validated, Knockdown Validated | |
| Q96EB6, Q923E4 | |
| SIRT1 | |
| A genomic peptide made to an internal region of the human SIRT1 protein (within residues 400-550). [Swiss-Prot Q96EB6] | |
| 0.1 mL | |
| Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Histone Deacetylases | |
| 23411 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction