Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 1/SIRT1 Antibody (PCRP-SIRT1-1E11), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314290R
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Sirtuin 1/SIRT1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-SIRT1-1E11 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
Recombinant full-length human Sirtuin 1/SIRT1protein (Uniprot: Q96EB6) | |
0.1 mL | |
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics | |
23411 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction