Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 1/SIRT1 Mouse anti-Human, Clone: PCRP-SIRT1-1E11, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Sirtuin 1/SIRT1 |
---|---|
Clone | PCRP-SIRT1-1E11 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array |
Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array |
Description
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
Sirtuin 1/SIRT1 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
Unconjugated | |
Mouse | |
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics | |
10 mM PBS with 0.05% BSA | |
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 | |
Recombinant full-length human Sirtuin 1/SIRT1protein (Uniprot: Q96EB6) | |
Primary | |
Store at 4°C. |
PCRP-SIRT1-1E11 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q96EB6 | |
23411 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title