Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 2/SIRT2 Antibody (PCRP-SIRT2-1A8), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308293B
Description
Sirtuin 2/SIRT2 Monoclonal antibody specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
Sirtuin 2/SIRT2 | |
Monoclonal | |
Biotin | |
EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
Recombinant full-length human Sirtuin 2/SIRT2 protein (Uniprot: Q8IXJ6 ) | |
100 μg | |
Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
22933 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-SIRT2-1A8 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction