Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 2/SIRT2 Mouse anti-Human, Clone: PCRP-SIRT2-1A8, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Sirtuin 2/SIRT2 |
---|---|
Clone | PCRP-SIRT2-1A8 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Description
Sirtuin 2/SIRT2 Monoclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
Sirtuin 2/SIRT2 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
22933 | |
IgG1 | |
Protein A or G purified |
PCRP-SIRT2-1A8 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2 | |
Recombinant full-length human Sirtuin 2/SIRT2 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title