Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 5/SIRT5 Antibody, Novus Biologicals™
SDP

Catalog No. p-5091149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056372T 0.025 mg
NB10056372 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056372T Supplier Novus Biologicals Supplier No. NB10056372SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Sirtuin 5/SIRT5 Polyclonal specifically detects Sirtuin 5/SIRT5 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Sirtuin 5/SIRT5
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NXA8
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ36950, NAD-dependent deacetylase sirtuin-5, silent mating type information regulation 2, S.cerevisiae, homolog 5, SIR2L5, sir2-like 5, SIR2-like protein 5, sirtuin (silent mating type information regulation 2 homolog) 5 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 5, sirtuin 5, sirtuin type 5
Gene Symbols SIRT5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 30-46 of human SIRT5 was used as immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23408
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.