Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sirtuin 5/SIRT5 Antibody (OTI2F1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274199B
Description
Sirtuin 5/SIRT5 Monoclonal specifically detects Sirtuin 5/SIRT5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
Sirtuin 5/SIRT5 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SIRT5 | |
Full length recombinant protein of human Sirtuin 5/SIRT5 (NP_036373) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
OTI2F1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
EC 3.5.1, EC 3.5.1.-, FLJ36950, NAD-dependent deacetylase sirtuin-5, silent mating type information regulation 2, S.cerevisiae, homolog 5, SIR2L5, sir2-like 5, SIR2-like protein 5, sirtuin (silent mating type information regulation 2 homolog) 5 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 5, sirtuin 5, sirtuin type 5 | |
Mouse | |
Affinity Purified | |
Epigenetics, Histone Deacetylases | |
23408.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction