Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SIS3
GREENER_CHOICE

Catalog No. 529150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-915-0 50 mg
52-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-915-0 Supplier Tocris Bioscience™ Supplier No. 5291/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Smad3 inhibitor; inhibits TGF-βR1 signaling

SIS3 is a selective Smad3 inhibitor; attenuates TGF-β1-dependent Smad3 phosphorylation and DNA binding. Has no effect on Smad2, p38 MAPK, ERK or PI 3-kinase signaling. Inhibits TGF-β1-induced myofibroblast differentiation of dermal fibroblasts. Also inhibits TGF-β2-induced endothelial cell differentiation in iPSCs.

Specifications

Quantity 50 mg
Product Type SIS3
Purity 97%
Molecular Weight (g/mol) 489.99

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.