Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIT Mouse anti-Human, Alexa Fluor 647, Clone: SIT-01 (also known as SIT-01), BD Pharmingen™

Catalog No. BDB568360 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568360 100 Tests
BDB568367 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB568360 Supplier BD Biosciences Supplier No. 568360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIT Monoclonal antibody specifically detects SIT in Human samples. It is validated for Intracellular staining (Flow Cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIT
Applications Flow Cytometry
Classification Monoclonal
Clone SIT-01 (also known as SIT-01)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias SHP-2 interacting transmembrane adaptor protein; SIT; SIT1; signaling threshold-regulating transmembrane adapter 1
Host Species Mouse
Immunogen Human Bacterially produced, recombinant, intracellular fragment of human SIT.
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.