Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SiteClick™ Antibody Labeling Kits

Catalog No. S10467
Encompass_Preferred
Click to view available options
Excitation/Emission:
405/525
405/565
405/585
405/605
405/625
405/655
405/705
496, 546, 565/578
Label or Dye:
Biotin
Qdot 525
Qdot 565
Qdot 585
Qdot 605
Qdot 625
Qdot 655
Qdot 705
Qdot 800
R-PE (R-Phycoerythrin)
Labeling Scale:
100 μg
100 to 250 μg

Generate brightly labeled antibodies for flow cytometry, fluorescence imaging, or western blot detection using SiteClick™ Qdot™ R-PE, and Biotin Antibody Labeling Kits.

Generate perfectly labeled antibodies with the SiteClick R-PE, Qdot, or Biotin Antibody Labeling Kits. Site selectivity is achieved by targeting the carbohydrate domains present on essentially all IgG antibodies, regardless of isotype and host species. Depending upon the label, the resulting SiteClick R-PE, Qdot, or Biotin conjugated antibody can be used in flow cytometry, fluorescence imaging, or Western blot detection. Each kit contains everything needed to label 100 μg of IgG antibody with an easy-to-follow, step-by-step protocol. Achieve highly efficient, site-specific, and reproducible labeling with the SiteClick R-PE, Qdot, or Biotin Antibody Labeling Kits.

Conventional amine or thiol labeling chemistry can result in labeling of the antigen binding domain or compromise the structure of an antibody. SiteClick labeling technology, however, attaches the label under mild conditions to the heavy chains of an IgG antibody, ensuring that the antigen binding domains remain available for binding to your antigen target. Because SiteClick Labeling Kits allow simple and gentle site-selective attachment of detection molecules to heavy chain N-linked glycans — far from the antigen-binding domain — excellent reproducibility is achieved during sequential labelings, as well from antibody to antibody. No harsh reduction steps are required, and antibody labeling is consistent and reproducible each time it is performed.

The SiteClick R-PE, Qdot, or Biotin Antibody labeling workflow consists of three steps:

1. antibody carbohydrate domain modification,
2. azide attachment to the antibody and
3. conjugation with the DIBO-modified label.

This workflow employs copper-free click chemistry to covalently link the label containing the DIBO moiety with the azide-modified antibody. Labeling antibodies with the SiteClick antibody labeling system gives you confidence that the antibody will be labeled the same way every time, with no need for reaction optimization. This means the same number of dyes are attached to each molecule, the antigen binding site is preserved, and the same results are achieved every time.

Important features of the SiteClick Qdot Antibody Labeling Kits:

• Include everything required to label 100 μg of IgG antibody (100–250 μg of IgG antibody with the SiteClick Biotin Antibody Labeling Kit)
• Easy to follow step-by-step protocol that require only a few hours hands-on time to complete.
• Highly efficient, site-specific, reproducible labeling chemistry results in high quality antibody conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Contains everything needed to azido modify and label 100 μg of antibody.
Product Type Labeling Kit
Quantity 1 kit
Content And Storage Store the entire kit at 2 to 6°C upon receipt
Excitation/Emission 496, 546, 565/578
Labeling Method SiteClick Labeling System
Labeling Scale 100 μg
Shipping Condition Wet Ice
Product Line SiteClick™
Label or Dye R-PE (R-Phycoerythrin)
Color Red-Orange
Labeling Target IgG
Flow Cytometer Laser Lines Blue, Green, Yellow
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.