Learn More
Invitrogen™ SiteClick™ Biotin Antibody Labeling Kit, 1 kit
Description
Achieve precise and reproducible antibody labeling with SiteClick Biotin Antibody Labeling Kits. This kit targets carbohydrate domains present on all IgG antibodies, regardless of isotype or host species, ensuring site-selective labeling. The labeled antibodies are suitable for flow cytometry, fluorescence imaging, and western blot detection.
Features of the SiteClick Biotin Antibody Labeling Kit include:
- Complete kit—includes everything needed to label 100–250 μg of IgG antibody using a straightforward, step-by-step protocol
- Site-specific labeling—SiteClick technology attaches labels to the heavy chains of IgG antibodies under mild conditions, preserving antigen binding domains
- Consistent results—the gentle, site-selective attachment to heavy-chain N-linked glycans ensures reproducibility across sequential labelings and different antibodies, without harsh reduction steps
- Efficient protocol—easy-to-follow steps requiring minimal hands-on time
- High-quality conjugates—produces highly efficient, site-specific, and reproducible antibody conjugates
Unlike traditional amine or thiol labeling, which can affect antigen binding domains or antibody structure, SiteClick labeling technology maintains the integrity of the antibody. This ensures that antigen binding sites remain available for target binding, providing consistent and reproducible labeling. The workflow utilizes copper-free click chemistry to covalently link the DIBO moiety with the azide-modified antibody, ensuring uniform labeling without the need for reaction optimization.
With the SiteClick Biotin Antibody Labeling Kit, you can confidently achieve consistent, high-quality labeling every time.
Order Info
Store in refrigerator (2–8°C), desiccate, protect from light, and do not freeze.
Specifications
Specifications
| Kit Contents | Contains everything needed to azido modify and label 100 μg of antibody |
| Product Type | Labeling Kit |
| Quantity | 1 kit |
| Content And Storage | Store the entire kit at 2 to 6°C upon receipt |
| Labeling Method | SiteClick Labeling System |
| Labeling Scale | 100 to 250 μg |
| Shipping Condition | Wet Ice |
| Product Line | SiteClick |
| Label or Dye | Biotin |
| Labeling Target | Antibodies |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.