Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIVA Antibody (OTI2F8), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274208AF532
Description
SIVA Monoclonal specifically detects SIVA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SIVA | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SIVA1 | |
Full length recombinant protein of human SIVA (NP_006418) produced in HEK293T cells. | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2F8 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CD27-binding (Siva) protein, CD27BPCD27-binding protein, Siva-1, SIVA1, apoptosis-inducing factor, Siva-2, SIVAapoptosis regulatory protein Siva | |
Mouse | |
0.1 mL | |
10572 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction