Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIVA Antibody, PerCP, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241150PCP
Description
SIVA Polyclonal antibody specifically detects SIVA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SIVA | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| CD27-binding (Siva) protein, CD27BPCD27-binding protein, Siva-1, SIVA1, apoptosis-inducing factor, Siva-2, SIVAapoptosis regulatory protein Siva | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SIVA. The immunogen is located within the first 50 amino acids of SIVA. Amino Acid Squence: AERYSQEVFEKTKRL | |
| 0.1 mL | |
| Apoptosis | |
| 10572 | |
| Store at 4C in the dark. | |
| IgY |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction