Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIX1 Antibody (OTI1H4), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274209AF488
Description
SIX1 Monoclonal specifically detects SIX1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| SIX1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SIX1 | |
| Recombinant protein fragment corresponding to amino acids 1-284 of human SIX1 (NP_005973) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1H4 | |
| Western Blot | |
| BOS3, deafness, autosomal dominant 23, DFNA23, homeobox protein SIX1, sine oculis homeobox (Drosophila) homolog 1, Sine oculis homeobox homolog 1, sine oculis homeobox homolog 1 (Drosophila), SIX homeobox 1, TIP39 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 6495.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction