Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551654 Supplier Invitrogen™ Supplier No. PA551654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CFKEKSRGVL REWYAHNPYP SPREKRELAE ATGLTTTQVS NWFKNRRQRD RAAEAKEREN TENNNSSSNK QNQLSPLEGG KPLMSSSEEE FSPPQSPDQN SVLLLQGNMG HARSSNYSLP GLTASQPSHG LQTHQHQLQD S Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal dominant deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIX1
Gene Accession No. Q15475
Gene Alias BB138287; BOS3; DFNA23; Homeobox protein SIX1; sine oculis homeobox homolog 1; sine oculis-related homeobox 1; sine oculis-related homeobox 1 homolog; SIX homeobox 1; SIX1; TIP39
Gene Symbols SIX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIX1. Recombinant protein control fragment (Product #RP-88902).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6495
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.