Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIX2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $648.50
Specifications
Antigen | SIX2 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SIX2 Polyclonal specifically detects SIX2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SIX2 | |
Polyclonal | |
Rabbit | |
Human | |
homeobox protein SIX2, sine oculis homeobox (Drosophila) homolog 2, Sine oculis homeobox homolog 2, sine oculis homeobox homolog 2 (Drosophila), SIX homeobox 2 | |
SIX2 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
10736 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ADPLQHHHGLQDSILNPMSANLVDLGS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title