Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541582 Supplier Invitrogen™ Supplier No. PA541582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WSLPACEHLH KNESVLKAKA VVAFHRGNFR ELYKILESHQ FSPHNHAKLQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%.

This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila 'sine oculis' homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SIX2
Gene Accession No. Q9NPC8
Gene Alias homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2; SIX2
Gene Symbols SIX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SIX2 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10736
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.