Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597149 Supplier Invitrogen™ Supplier No. PA597149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIWDGEETSY CFKEKSRSVL REWYAHNPYP SPREKRELAE ATGLTTTQVS NWFKN Positive Samples: Rat kidney; Cellular Location: Nucleus.

This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila 'sine oculis' homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SIX2
Gene Accession No. Q9NPC8
Gene Alias homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2; SIX2
Gene Symbols SIX2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-174 of human SIX2 (NP_0586283).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10736, 366542
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.