Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119714
This item is not returnable.
View return policy
Description
Positive Control: Mouse kidney tissue lysate, mouse stomach tissue lysate. Subcellular Location: Nucleus.
This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila 'sine oculis' homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
Specifications
SIX2 | |
Polyclonal | |
Unconjugated | |
SIX2 | |
homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2; SIX2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20472 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q62232 | |
SIX2 | |
Synthetic peptide within mouse six2 aa 100-160. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction