Learn More
SIX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115621AP150UL

Description
This antibody is a rabbit polyclonal antibody raised against full length SIX2 of human origin.
This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila ′sine oculis′ homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.Specifications
SIX2 | |
Polyclonal | |
Unconjugated | |
SIX2 | |
Homeobox protein SIX2, Sine oculis homeobox homolog 2, SIX homeobox 2, SIX2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10736, 20472, 366542 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62232, Q9NPC8 | |
Six2 | |
SIX2 Fusion Protein Ag2124 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.