Learn More
Invitrogen™ SIX3 Monoclonal Antibody (4H6)

Description
Peptide Sequence: QAIGPSGMRS LAEPGCPTHG SAESPSTAAS PTTSVSSLTE RADTGTSILS VTSSDSECDV.
Specifications
Specifications
| Antigen | SIX3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4H6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Six3 |
| Gene Accession No. | O95343 |
| Gene Alias | E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.