Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540530 Supplier Invitrogen™ Supplier No. PA540530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VFRSPLDLYS SHFLLPNFAD SHHRSILLAS SGGGNGAGGG GGAGGGSGGG Sequence homology: Cow: 93%; Dog: 86%; Human: 100%; Mouse: 86%; Rat: 86%.

This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIX3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Six3
Gene Accession No. O95343
Gene Alias E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta
Gene Symbols Six3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SIX3 (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6496
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.