Learn More
Invitrogen™ SIX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113637
Description
Immunogen sequence: MRSLAEPGCP THGSAESPST AASPTTSVSS LTERADTGTS ILSVTSSDSE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.
This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.
Specifications
SIX3 | |
Polyclonal | |
Unconjugated | |
Six3 | |
E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20473 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q62233 | |
Six3 | |
Synthetic peptide directed towards the C terminal region of mouse Six3 (aa 281-330). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.