Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX3 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990494
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0494 50 μg
50-105-7532 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0494 Supplier Rockland Immunochemicals Supplier No. 200201A26
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with mouse Six3. Cross-reactivity with Six3 from other sources has not been determined.

This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.

Specifications

Antigen SIX3
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Six3
Gene Accession No. Q62233
Gene Alias BOS_11581; E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; Sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta
Gene Symbols Six3
Host Species Guinea Pig
Immunogen This Protein A purified antibody was prepared from whole guinea pig serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of mouse Six3 protein.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20473
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.